EMAIL THIS PAGE TO A FRIEND
Current protocols in cell biology
Fluorescent staining of subcellular organelles: ER, Golgi complex, and mitochondria.
PMID 18228364
Abstract
The ability to distinguish and identify specific subcellular compartments is essential to understanding organelle function, biogenesis, and maintenance within cells and to defining protein trafficking pathways. Fluorescent dyes and/or fluorescently labeled lipid derivatives can be used to identify ER, Golgi complex, and mitochondria. Specific conditions for labeling each of these compartments are described.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
318426
3,3′-Dihexyloxacarbocyanine iodide, 98%
C29H37IN2O2
N8278
N-(NBD-Aminohexanoyl)sphingosine
C30H49N5O6