EMAIL THIS PAGE TO A FRIEND
Wiadomosci parazytologiczne
[Experimental studies of the control of mosquitoes of the genus Anopheles using selected insecticides].
PMID 1823486
Abstract
In the research 4 compounds: Iodofenphos, Propetamphos, Dioxacarb and Permethrin and two commercial insecticides: Pibutoks Super (Permethrin) and Safrotin 10 WP (Propetamphos) were used to control anophelini females. The chemicals were tested in the laboratory on mosquitoes caught in the nature and the contact method was employed. Propetamphos and Permethrin at concentrations 0.05% and 0.1%, after 2 hours of 1 hour exposure reduced the mosquitoes in 96% and 99.9% respectively. Iodofenphos and Dioxacarb, even at concentration 0.1%, after 24 hours of 1 hour exposure caused barely 78.3% and 83.9% mortality of these insects. Both insecticides tested, in the 7 days period after the spraying, caused more than 97% reduction of the Anopheles population in the cow-sheds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45544
Jodfenphos, PESTANAL®, analytical standard
C8H8Cl2IO3PS
34371
Propetamphos, PESTANAL®, analytical standard
C10H20NO4PS