EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Iron-catalyzed asymmetric olefin cis-dihydroxylation with 97% enantiomeric excess.
PMID 18236485
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
712337
(2R,2′R)-1,1′-Bis(6-methyl-2-pyridinylmethyl)-2,2′-bipyrrolidine tetrahydrochloride, 96%
C22H30N4 · 4HCl