EMAIL THIS PAGE TO A FRIEND
Vector borne and zoonotic diseases (Larchmont, N.Y.)
Human infection with Moniliformis moniliformis (Bremser 1811) (Travassos 1915) in Iran: another case report after three decades.
PMID 18237263
Abstract
This article reports on the occurrence of M. moniliformis in a 2-year-old girl in Iran after a long time period from its last recorded occurrence. Diagnosis of the both the earlier human record, as well as the later one that we are discussing in this report, has not initially been established by routine stool examination. In this case, symptoms that may suggest small bowel disease or dysfunction were not significantly seen. The worm obtained from the child was identified as a female M. moniliformis, but how the child became infected is unknown.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46632
Citric acid sesquipiperazine salt hydrate, VETRANAL®, analytical standard
C6H8O7 · 1.5C4H10N2 · xH2O