EMAIL THIS PAGE TO A FRIEND
IEEE transactions on ultrasonics, ferroelectrics, and frequency control
Piezoelectric sensor applied in ultrasonic contact microscopy for the investigation of material surfaces.
PMID 18238418
Abstract
In this paper the theoretical analysis and the results of testing of a piezoelectric cantilever for the investigation of material surfaces are presented. The cantilever consists of a thin piezoelectric plate bonded with a thin metal (e.g., molybdenum) foil. The analytical formulae for spring constants and sensitivity of such monomorph have been established. The performed analysis permits us to state the optimal parameters of the sensor dimensions and the Young's modulus of applied materials. An important factor is the position of the nodal plane and its influence on sensor sensitivity. Various combinations of ceramic plates and metal foils were theoretically analyzed, then applied in practical realizations of the sensors. The sensor with a tip was applied to a contact ultrasonic microscope, and replaced the optical sensor commonly used to measure the deflection amplitude of the tip during scanning the surfaces of the sample. Such a method enables us to create the three-dimensional images of the surface. The sensors produced were calibrated using the quasi-static method. Sensors of this kind may fulfil requirements necessary in investigating surface properties of materials applied in modern electronics and technology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF42672427
Molybdenum, foil, not light tested, 25x25mm, thickness 0.003mm, 99.9%
Mo
GF11484750
Molybdenum, foil, not light tested, 25x25mm, thickness 0.01mm, 99.9%
Mo
GF81739457
Molybdenum, foil, not light tested, 50x50mm, thickness 0.003mm, 99.9%
Mo
GF30193781
Molybdenum, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.9%
Mo
GF66431606
Molybdenum, foil, not light tested, 25x25mm, thickness 0.006mm, 99.9%
Mo
GF23412383
Molybdenum, foil, not light tested, 25x25mm, thickness 0.008mm, 99.9%
Mo
GF90888788
Molybdenum, foil, not light tested, 25x25mm, thickness 0.0125mm, 99.9%
Mo
GF98811355
Molybdenum, foil, not light tested, 25x25mm, thickness 0.01mm, annealed, 99.9%
Mo
GF26098027
Molybdenum, foil, not light tested, 50x50mm, thickness 0.001mm, 99.9%
Mo
GF63646218
Molybdenum, foil, not light tested, 50x50mm, thickness 0.0025mm, 99.9%
Mo
GF20045786
Molybdenum, foil, not light tested, 50x50mm, thickness 0.002mm, 99.9%
Mo
GF47495975
Molybdenum, foil, not light tested, 50x50mm, thickness 0.004mm, 99.9%
Mo
GF48792628
Molybdenum, foil, not light tested, 50x50mm, thickness 0.005mm, 99.9%
Mo
GF74350267
Molybdenum, foil, not light tested, 50x50mm, thickness 0.006mm, 99.9%
Mo
GF37145358
Molybdenum, foil, not light tested, 50x50mm, thickness 0.007mm, 99.9%
Mo
GF95631419
Molybdenum, foil, not light tested, 50x50mm, thickness 0.008mm, 99.9%
Mo
GF41758768
Molybdenum, foil, not light tested, 50x50mm, thickness 0.009mm, 99.9%
Mo
GF99294968
Molybdenum, foil, not light tested, 50x50mm, thickness 0.01mm, 99.9%
Mo
GF42252016
Molybdenum, foil, not light tested, 50x50mm, thickness 0.01mm, annealed, 99.9%
Mo
GF61564552
Molybdenum, foil, 100x100mm, thickness 0.5mm, annealed, 99.9%
Mo
GF91334213
Molybdenum, foil, 100x100mm, thickness 2.0mm, annealed, 99.9%
Mo
GF69690291
Molybdenum, foil, 150x150mm, thickness 0.25mm, annealed, 99.9%
Mo
GF60494799
Molybdenum, foil, 250x300mm, thickness 0.1mm, annealed, 99.9%
Mo
GF79159023
Molybdenum, foil, 25x25mm, thickness 0.30mm, annealed, 99.9%
Mo
GF11286345
Molybdenum, foil, 300x300mm, thickness 0.25mm, annealed, 99.9%
Mo
GF04107620
Molybdenum, foil, 50x50mm, thickness 0.20mm, annealed, 99.9%
Mo
GF09623471
Molybdenum, foil, 50x50mm, thickness 0.75mm, annealed, 99.9%
Mo
GF52486780
Molybdenum, foil, 0.2m coil, thickness 0.0125mm, 99.9%
Mo
GF72844386
Molybdenum, foil, 4mm disks, thickness 0.01mm, annealed, 99.9%
Mo
GF94854099
Molybdenum, foil, 4mm disks, thickness 0.125mm, annealed, 99.9%
Mo