EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Evaluation of a series of bicyclic CXCR2 antagonists.
PMID 18240390
Abstract
The CXCR2 SAR of a series of bicyclic antagonists such as the 2-aminothiazolo[4,5-d]pyrimidine 3b was investigated by systematic variation of the fused pyrimidine-based heterocyclic cores. Replacement of the aminothiazole ring with a 2-thiazolone alternative led to a series of thiazolo[4,5-d]pyrimidine-2(3H)-one antagonists with markedly improved biological and pharmacokinetic properties, which are suitable pharmacological tools to probe the in vivo effects of CXCR2 antagonism combined with the associated CCR2 activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
265314
2,3-Difluorobenzyl bromide, 98%
C7H5BrF2