EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis of new 3-pyridinecarboxylates of potential vasodilation properties.
PMID 18241958
Abstract
2-(alicyclic-amino)-4,6-diaryl-3-pyridinecarboxylates 5a-d were prepared via aromatic nucleophilic substitution reaction of secondary amines (piperidine or morpholine) with 2-bromo-3-pyridinecarboxylate derivatives 3a,b. The latters were obtained through bromination of 3-aryl-4-benzoyl-2-cyanobutyrates 2a and 2b, which were obtained from the base promoted addition of ethyl cyanoacetate to 2-propen-1-ones 1a and 1b, with bromine in glacial acetic acid. Reaction of 3 with piperazine hexahydrate in 2:1 molar ratio afforded 1,4-bis[(ethyl 4,6-diaryl-3-pyridinecarboxylate)-2-yl]piperazines 6a,b. Reaction of 3 with anilines in refluxing pyridine unexpectedly gave 2-(aryl-amino)-3-pyridinecarboxylates 8a-g and 2-amino-3-pyridinecarboxylates 9a and 9b. Vasodilation activity screening for the synthesized pyridinecarboxylates using isolated thoracic aortic rings' standard method of rats shows considerable properties. Compounds 5b, 5c, 6b and 8g reveal remarkable vasodilation potency (IC50, concentrations necessary for 50% reduction of maximal norepinephrine hydrochloride induced contracture) 0.175, 0.146, 0.229 and 0.233 mM, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E40609
Ethyl nicotinate, 99%
C8H9NO2