EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Solubility enhancement of gluten and organic compounds by arginine.
PMID 18242019
Abstract
Arginine suppresses protein-protein and protein-surface interactions and thus is expected to increase the solubility of the proteins. We have examined here the effects of arginine on the solubility of a highly insoluble protein, gluten, and two organic compounds, octyl-gallate and coumarin, which have low to moderate aqueous solubilities. Arginine significantly increased the solubility of these molecules concentration dependently, while a weak salting-out salt, NaCl, decreased it. The observed ability of arginine to salt-in these compounds can be explained from its binding to aromatic groups and protein surface. Such solubilizing action of arginine may be used to enhance the solubility of poorly soluble organic drug substances.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48700
Octyl gallate, antioxidant, ≥99.0% (HPLC)
C15H22O5
06717
Octyl gallate, analytical standard
C15H22O5