EMAIL THIS PAGE TO A FRIEND
Clinical biochemistry
Is secondary wavelength always necessary in turbidimetric urine protein measurements?
PMID 18249194
Abstract
Turbidimetric urine protein methods like Benzethonium Chloride (BTC) and Benzalkonium Chloride (BC) are previously reported to give falsely low protein results in very high protein concentrations (>2000 mg/L). BTC and BC assays were adapted to an Olympus AU 640 analyzer in order to investigate the effect of the secondary wavelength on the linearity of the assays. Pyrogallol red (PYR) was also adapted to the same analyzer and used as the comparative method. The linearity of BC was increased from 1500 mg/L to 3000 mg/L and the linearity of BTC was increased from 1500 mg/L to 2500 mg/L with the omission of the secondary wavelength. We propose single-wavelength measurements for BC and BTC methods; however, urine samples with protein concentration >2500 mg/L for BTC and >3000 mg/L for BC should be diluted to appropriate concentrations.