EMAIL THIS PAGE TO A FRIEND
Journal of periodontal research
Biochemical comparison of proteolytic enzymes present in rough- and smooth-surfaced capnocytophagas isolated from the subgingival plaque of periodontitis patients.
PMID 1825330
Abstract
Four rough-surfaced (R) and three smooth-surfaced (S) clinical isolates of Capnocytophaga obtained from the subgingival plaque of periodontitis patients were studied for their peptidase and protease profiles. The results were compared with those obtained with C. gingivalis (which has a smooth morphology). All cell extracts obtained by ultrasonic treatment displayed high peptidase activity toward N-aminoacyl-2-naphthylamines, the best substrates being the arginyl, aspartyl, and leucyl derivatives. The R and S isolates did not differ in these enzyme activities. Also the protease profiles studies with 4-phenylazobenzyloxycarbonyl-L-prolyl-L-leucylglycyl-L-proly l-D-arginine (PZ-PLPGA) and casein were similar. All extracts also hydrolyzed furylacryloyl-L-leucylglycyl-L-prolyl-L-alanine (FALGPA), reconstituted type I [3H]-collagen, and gelatin. N alpha-Benzoyl-DL-rginyl-2-naphthylamine was hydrolyzed faster by the R than the S strains. Comparison between cell suspensions and cell extracts of C. gingivalis showed the suspensions to be enzymatically more active than the extracts. In general, peptidase substrates and PZ-PLGPA were hydrolyzed at a higher rate by suspensions than by extracts, while protease substrates (such as casein) were hydrolyzed faster by the extracts. Gelatin and FALGPA were hydrolyzed by cell extracts only. Fast protein liquid chromatography of peptidases on a gel column was found to be a suitable method to differentiate between R and S isolates in diagnostics, while the chromatographic profiles of proteases were not suitable for this purpose.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5135
N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala
C23H32N4O7