EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
Beta-eudesmol suppresses tumour growth through inhibition of tumour neovascularisation and tumour cell proliferation.
PMID 18253884
Abstract
In the present study, we investigated the potential anti-angiogenic mechanism and anti-tumour activity of beta-eudesmol using in vitro and in vivo experimental models. Proliferation of human umbilical vein endothelial cells (HUVEC) stimulated with vascular endothelial growth factor (VEGF, 30 ng/ml) and basic fibroblast growth factor (bFGF, 30 ng/ml) was significantly inhibited by beta-eudesmol (50-100 microM). Beta-eudesmol (100 microM) also blocked the phosphorylation of cAMP response element binding protein (CREB) induced by VEGF (30 ng/ml) in HUVEC. Beta-eudesmol (10-100 microM) inhibited proliferation of HeLa, SGC-7901, and BEL-7402 tumour cells in a time- and dose-dependent manner. Moreover, beta-eudesmol treatment (2.5-5 mg/kg) significantly inhibited growth of H(22) and S(180) mouse tumour in vivo. These results indicated that beta-eudesmol inhibited angiogenesis by suppressing CREB activation in growth factor signalling pathway. This is the first study to demonstrate that beta-eudesmol is an inhibitor of tumour growth.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17790
β-Eudesmol, ≥90% (GC)
C15H26O