EMAIL THIS PAGE TO A FRIEND
Advances in prostaglandin, thromboxane, and leukotriene research
Opposite effects of PGI2 and PAF generated during acute myocardial ischemia in the perfused rabbit heart.
PMID 1825437
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4904
β-Acetyl-γ-O-hexadecyl-L-α-phosphatidylcholine hydrate, ≥98%
C26H54NO7P · xH2O