EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and biological evaluation of novel 8-aminomethylated oroxylin A analogues as alpha-glucosidase inhibitors.
PMID 18255289
Abstract
A series of 8-aminomethylated derivatives (1a-1j) were prepared by Mannich reaction of oroxylin A (1) with appropriate primary or secondary amines and para-formaldehyde. All the compounds were tested for their alpha-glucosidase inhibition activity against both yeast and rat intestinal alpha-glucosidase. Some of the compounds demonstrated significantly better alpha-glucosidase inhibitory activity than the parent compound (oroxylin A).