Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Interaction of pepsin with aromatic amino acids and their derivatives immobilized to Sepharose.
PMID 18255363
The interaction of porcine pepsin A with immobilized derivatives of aromatic amino acids was investigated. Divinyl sulfone-activated Sepharose was used to immobilize N-acetyl-l-phenylalanine and 3,5-diiodo-l-tyrosine via their free carboxyl groups and l-tyrosine via its amino group. Immobilized l-tyrosine was iodinated after coupling. The optimum conditions for the separation of porcine pepsin A using the prepared affinity carriers were studied and the following parameters were established: enzyme recovery, reproducibility of analyses, capacity and dependence of the elution peak area on the concentration of the loaded enzyme. The ability of the prepared affinity carriers to retain various types of proteins was compared under optimum conditions for porcine pepsin A separation. While immobilized 3,5-diiodo-l-tyrosine and iodinated l-tyrosine-Sepharose adsorbed relatively high amounts of bovine serum albumin and ovalbumin, only negligible amounts of these proteins were adsorbed to immobilized N-acetyl-l-phenylalanine. The behavior of porcine pepsin A was the same as its complex with pepstatin A on the prepared affinity carriers, indicating that the enzyme active site is not involved in the studied interaction.
Related Materials
Product #
Molecular Formula
Add to Cart
3,5-Diiodo-L-tyrosine dihydrate, crystalline
C9H9I2NO3 · 2H2O