EMAIL THIS PAGE TO A FRIEND
Se pu = Chinese journal of chromatography
[Determination of balofloxacin in human plasma by HPLC with solid-phase extraction].
PMID 18257322
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32153
Balofloxacin, VETRANAL, analytical standard
C20H24FN3O4