EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Synthesis and evaluation of a novel ligand for folate-mediated targeting liposomes.
PMID 18258394
Abstract
Folate receptors (FRs) have been identified as cellular surface markers for cancer and leukemia. Liposomes containing lipophilic derivatives of folate have been shown to effectively target FR-expressing cells. Here, we report the synthesis of a novel lipophilic folate derivative, folate-polyethylene glycol-cholesterol hemisuccinate (F-PEG-CHEMS), and its evaluation as a targeting ligand for liposomal doxorubicin (L-DOX) in FR-expressing cells. Liposomes containing F-PEG-CHEMS, with a mean diameter of 120+/-20 nm, were synthesized by polycarbonate membrane extrusion and were shown to have excellent colloidal stability. The liposomes were taken up selectively by KB cells, which overexpress FR-alpha. Compared to folate-PEG-cholesterol (F-PEG-Chol), which contains a carbamate linkage, F-PEG-CHEMS better retained its FR-targeting activity during prolonged storage. In addition, F-PEG-CHEMS containing liposomes loaded with DOX (F-L-DOX) showed greater cytotoxicity (IC(50)=10.0muM) than non-targeted control L-DOX (IC(50)=57.5 microM) in KB cells. In ICR mice, both targeted and non-targeted liposomes exhibited long circulation properties, although F-L-DOX (t(1/2)=12.34 h) showed more rapid plasma clearance than L-DOX (t(1/2)=17.10h). These results suggest that F-PEG-CHEMS is effective as a novel ligand for the synthesis of FR-targeted liposomes.