EMAIL THIS PAGE TO A FRIEND
Arteriosclerosis, thrombosis, and vascular biology
High glucose enhances transient receptor potential channel canonical type 6-dependent calcium influx in human platelets via phosphatidylinositol 3-kinase-dependent pathway.
PMID 18258814
Abstract
Transient receptor potential canonical type 6 (TRPC6) channels mediating 1-oleoyl-2-acetyl-sn-glycerol (OAG)-induced calcium entry have been identified on human platelets. In the present study we tested the hypothesis that hyperglycemia increases the expression of TRPC6 channels. Platelets from healthy control subjects and patients with type 2 diabetes mellitus were incubated with glucose and calcium influx was measured using the fluorescent dye technique. TRPC channel protein expression was investigated using immunofluorescence and fluorescence microscopy of single platelets. Administration of 25 mmol/L glucose significantly enhanced the OAG-induced calcium influx, which was attenuated by inhibitors of the phosphatidylinositol 3-kinase, wortmannin or LY294002. The glucose-enhanced and OAG-induced calcium influx was concentration- and time-dependent. Glucose significantly increased the TRPC6 protein expression in platelets to 131+/-12% (n=33; P<0.05), whereas the expression of TRPC1, TRPC3, TRPC4, or TRPC5 were unchanged. The glucose-induced TRPC6 expression was significantly attenuated in the presence of wortmannin or LY294002. Platelets from patients with type 2 diabetes mellitus showed increased TRPC6 expression compared to nondiabetic individuals (P<0.05). The study indicates that high glucose increases TRPC6 channel protein expression on the platelet surface which is mediated by a phosphatidylinositol 3-kinase-dependent pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O6754
1-Oleoyl-2-acetyl-sn-glycerol, ≥97% (TLC), oil
C23H42O5