EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Electrophoresis PDMS/glass chips with continuous on-chip derivatization and analysis of amino acids using naphthalene-2,3-dicarboxaldehyde as fluorogenic agent.
PMID 18261517
Abstract
In this work, we developed a PDMS electrophoresis device able to carry out on-chip derivatization and quantification of amino acids (AAs) using naphthalene-2,3-dicarboxaldehyde (NDA) as a fluorogenic agent. A chemical modification of the PDMS surface was found compulsory to achieve the derivatization of AAs with NDA and a limit of detection (LOD) of 40nM was reached for glycine. Finally, we suggested the applicability of this microdevice for the analysis of real biological samples such as a rat hippocampus microdialysate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
70215
2,3-Naphthalenedicarboxaldehyde, suitable for fluorescence
C12H8O2