EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis of eudistomin D analogues and its effects on adenosine receptors.
PMID 18262425
Abstract
Six analogues (1-6) of eudistomin D, a beta-carboline alkaloid from a marine tunicate Eudistoma olivaceum, were synthesized, and their affinity and selectivity for adenosine receptors A(1), A(2A), and A(3) were examined. All the synthetic compounds 1-6 did not show affinity to the adenosine A(1) receptor. Delta-carboline 3 exhibited the most potent affinity to the adenosine receptor A(3) among compounds 1-6. Delta-carbolines 3 and 4 showed better affinity than the corresponding beta-carbolines 1 and 2, respectively, while N-methylation (2, 4, and 6, respectively) of the pyrrole ring in 1, 3, and 5 resulted in the reduced affinity to the adenosine A(3) receptor. On the other hand, an eudistomin D derivative, BED, exhibited modest affinity to all the receptors A(1), A(2A), and A(3) but no selectivity.