EMAIL THIS PAGE TO A FRIEND
Journal of separation science
Simultaneous determination by UPLC-ESI-MS of scoparone, capillarisin, rhein, and emodin in rat urine after oral administration of Yin Chen Hao Tang preparation.
PMID 18264991
Abstract
A simple, sensitive, and validated method was developed for simultaneous determination of scoparone, capillarisin, rhein, and emodin in rat urine by ultra-performance liquid chromatography/electrospray ionization quadruple time-of-flight mass spectrometry (UPLC-MS). The urinary samples were analyzed on an Acquity UPLC BEH C18 1.7 microm 2.1x50 mm column. Scoparone, capillarisin, rhein, and emodin in rat urine were simultaneously analyzed with good separation. The lower limits of detection were 6.0, 9.0, 7.0, and 3.0 ng/mL, and the lower limits of quantification were 20.0, 33.0, 24.0, and 12.0 ng/mL for scoparone, capillarisin, rhein, and emodin, respectively. The intra- and inter-day precisions (RSD) were less than 9%. The intra- and inter-accuracies were found to be in the range of 94.14-104.54% for scoparone, 101.72-107.34% for capillarisin, 95.24-103.59% for rhein, and 101.32-107.82% for emodin at three concentration levels. The absolute recoveries for scoparone, capillarisin, rhein, and emodin were not less than 77.0%. The developed method has been applied to determine scoparone, capillarisin, rhein, and emodin in rat urine after oral administration of Yin Chen Hao Tang preparation, a traditional Chinese medicine formulation widely used in China for treatment of jaundice and liver disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
254886
6,7-Dimethoxycoumarin, 98%
C11H10O4