EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
A chemical method for fast and sensitive detection of DNA synthesis in vivo.
PMID 18272492
Abstract
We have developed a method to detect DNA synthesis in proliferating cells, based on the incorporation of 5-ethynyl-2'-deoxyuridine (EdU) and its subsequent detection by a fluorescent azide through a Cu(I)-catalyzed [3 + 2] cycloaddition reaction ("click" chemistry). Detection of the EdU label is highly sensitive and can be accomplished in minutes. The small size of the fluorescent azides used for detection results in a high degree of specimen penetration, allowing the staining of whole-mount preparations of large tissue and organ explants. In contrast to BrdU, the method does not require sample fixation or DNA denaturation and permits good structural preservation. We demonstrate the use of the method in cultured cells and in the intestine and brain of whole animals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
900584
5-Ethynyl-2′-deoxyuridine, 95%
C11H12N2O5
BCK488-IV-FC-L
In Vivo EdU Flow Cytometry 50 Kit 488, sufficient for 50 assays
BCK488-IV-FC-M
In Vivo EdU Flow Cytometry 50 Kit 488, sufficient for 50 assays
BCK488-IV-FC-S
In Vivo EdU Flow Cytometry 50 Kit 488, sufficient for 50 assays
BCK555-IV-FC-L
In Vivo EdU Flow Cytometry Kit 555, sufficient for 50 assays
BCK555-IV-FC-M
In Vivo EdU Flow Cytometry Kit 555, sufficient for 50 assays
BCK555-IV-FC-S
In Vivo EdU Flow Cytometry Kit 555, sufficient for 50 assays
BCK594-IV-FC-L
In Vivo EdU Flow Cytometry Kit 594, sufficient for 50 assays
BCK594-IV-FC-M
In Vivo EdU Flow Cytometry Kit 594, sufficient for 50 assays
BCK594-IV-FC-S
In Vivo EdU Flow Cytometry Kit 594, sufficient for 50 assays
BCK647-IV-FC-L
In Vivo EdU Flow Cytometry Kit 647, sufficient for 50 assays
BCK647-IV-FC-M
In Vivo EdU Flow Cytometry Kit 647, sufficient for 50 assays
BCK647-IV-FC-S
In Vivo EdU Flow Cytometry Kit 647, sufficient for 50 assays
BCK488-IV-HTS-L
In Vivo EdU HTS 2x96 Kit 488, sufficient for 200 assays
BCK488-IV-HTS-M
In Vivo EdU HTS 2x96 Kit 488, sufficient for 200 assays
BCK488-IV-HTS-S
In Vivo EdU HTS 2x96 Kit 488, sufficient for 200 assays
BCK555-IV-HTS-L
In Vivo EdU HTS 2x96 Kit 555, sufficient for 200 assays
BCK555-IV-HTS-M
In Vivo EdU HTS 2x96 Kit 555, sufficient for 200 assays
BCK555-IV-HTS-S
In Vivo EdU HTS 2x96 Kit 555, sufficient for 200 assays