EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V
Commentary to the article "Human skin penetration of the major components of Australian tea tree oil applied in its pure form and as a 20% solution in vitro".
PMID 18280124
Abstract
This note summarises recent studies on skin penetration of terpinen-4-ol, which is the main component of tea tree oil [S.E. Cross, M. Russell, I. Southwell, M.S. Roberts, Human skin penetration of the major components of Australian tea tree oil applied in its pure form and as a 20% solution in vitro, Eur. J. Pharm. Biopharm. doi: 10.1016/j.ejpb.2007.10.002 (in press)]. The influence of different experimental models on obtained skin penetration results is discussed.