EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
The Ehrlich pathway for fusel alcohol production: a century of research on Saccharomyces cerevisiae metabolism.
PMID 18281432
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W249715
Fusel oil, natural, FG