EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A new, simple approach to confer permanent antimicrobial properties to hydroxylated surfaces by surface functionalization.
PMID 18283346
Abstract
A new, simple method to obtain ultrathin polycationic monolayers on hydroxylated surfaces is described which uses a bifunctional copolymer comprising a reactive part (trimethoxysilane) and positive charges (quaternary ammonium salts) to confer antimicrobial properties.