EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Prevalence and susceptibility profile of Candida metapsilosis and Candida orthopsilosis: results from population-based surveillance of candidemia in Spain.
PMID 18285486
Abstract
We describe the prevalences and susceptibility profiles of two recently described species, Candida metapsilosis and Candida orthopsilosis, related to Candida parapsilosis in candidemia. The prevalences of these species (1.7% for C. metapsilosis and 1.4% for C. orthopsilosis) are significant. Differences observed in their susceptibility profiles could have therapeutic importance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F7129
5-Fluorocytosine, nucleoside analog
C4H4FN3O