EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
CD3 stimulation causes phosphorylation of phospholipase C-gamma 1 on serine and tyrosine residues in a human T-cell line.
PMID 1828897