EMAIL THIS PAGE TO A FRIEND
Chemistry & biology
Molecular analysis of the kirromycin biosynthetic gene cluster revealed beta-alanine as precursor of the pyridone moiety.
PMID 18291322
Abstract
Kirromycin is a complex linear polyketide that acts as a protein biosynthesis inhibitor by binding to the bacterial elongation factor Tu. The kirromycin biosynthetic gene cluster was isolated from the producer, Streptomyces collinus Tü 365, and confirmed by targeted disruption of essential biosynthesis genes. Kirromycin is synthesized by a large hybrid polyketide synthase (PKS)/nonribosomal peptide synthetase (NRPS) encoded by the genes kirAI-kirAVI. This complex involves some very unusual features, including the absence of internal acyltransferase (AT) domains in KirAI-KirAV, multiple split-ups of PKS modules on separate genes, and swapping in the domain organization. Interestingly, one PKS enzyme, KirAVI, contains internal AT domains. Based on in silico analysis, a route to pyridone formation involving PKS and NRPS steps was postulated. This hypothesis was experimentally proven by feeding studies with [U-13C3(15)N]beta-alanine and NMR and MS analyses of the isolated pure kirromycin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K1507
Kirromycin from Streptomyces collinus
C43H60N2O12