EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Development of novobiocin analogues that manifest anti-proliferative activity against several cancer cell lines.
PMID 18293999
Abstract
Recent studies have shown that the DNA gyrase inhibitor, novobiocin, binds to a previously unrecognized ATP-binding site located at the C-terminus of Hsp90 and induces degradation of Hsp90-dependent client proteins at approximately 700 microM. As a result of these studies, several analogues of the coumarin family of antibiotics have been reported and shown to exhibit increased Hsp90 inhibitory activity; however, the monomeric species lacked the ability to manifest anti-proliferative activity against cancer cell lines at concentrations tested. In an effort to develop more efficacious compounds that produce growth inhibitory activity against cancer cell lines, structure-activity relationships were investigated surrounding the prenylated benzamide side chain of the natural product. Results obtained from these studies have produced the first novobiocin analogues that manifest anti-proliferative activity against several cancer cell lines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
766593
Potassium 1-thianthrenyltrifluoroborate, 90%
C12H7BF3KS2