EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Carbonic anhydrase inhibitors: the very weak inhibitors dithiothreitol, beta-mercaptoethanol, tris(carboxyethyl)phosphine and threitol interfere with the binding of sulfonamides to isozymes II and IX.
PMID 18295485
Abstract
The inhibition of the metalloenzyme carbonic anhydrase (CA, EC 4.2.1.1) with dithiothreitol, 2-mercaptoethanol, tris(carboxyethyl)phosphine (reducing agent frequently added to enzyme assay buffers) and threitol has been investigated. The agents were very weak inhibitors of isozymes CA II and CA IX, but unexpectedly, strongly influenced the binding of the low nanomolar sulfonamide inhibitor acetazolamide (5-acetamido-1,3,4-thiadiazole-2-sulfonamide). Acetazolamide affinity for all investigated CAs diminished orders of magnitude with increasing concentrations of these agents in the assay system. DTT and similar derivatives should not be added to the assay buffers used in monitoring CA activity/inhibition, as they lead to under-estimation of the binding constants, by a mechanism probably involving the formation of ternary complexes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
377619
D-Threitol, 99%
C4H10O4
263559
DL-Threitol, 97%
C4H10O4