EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
From furans to anilines: toward one-pot two-step amination/diels-alder sequences.
PMID 18302410
Abstract
Selective metal-free amination and Diels-Alder reactions are described in the furan series, leading to polysubstituted anilines or to stable oxabicyclic adducts in high yield. Interestingly, anilines are conveniently prepared through a novel one-pot, two-step amination/Diels-Alder procedure from commercially available 5-bromo-2-furaldehyde.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
433985
5-Bromo-2-furaldehyde, 97%
C5H3BrO2