EMAIL THIS PAGE TO A FRIEND
Organic letters
Efficient enantioselective hetero-Diels-Alder reaction of Brassard's diene with aliphatic aldehydes: a one-step synthesis of (R)-(+)-kavain and (S)-(+)-dihydrokavain.
PMID 18303910
Abstract
An efficient catalytic asymmetric hetero-Diels-Alder reaction of Brassard's diene with aliphatic aldehydes was reported. The catalyst, which was generated from (R)-BINOL, Ti(i-PrO)4, and 4-picolyl chloride hydrochloride, promoted the reaction smoothly to afford the corresponding alpha,beta-unsaturated delta-lactone derivatives in moderate-to-good yields (46-79%) with high enantioselectivities (up to 88% ee). Natural products (R)-(+)-kavain (70% ee, >99% ee after single recrystallization) and (S)-(+)-dihydrokavain (84% ee) were also prepared in one step by using this methodology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
41866
(S)-(+)-7,8-Dihydrokavain, analytical standard
C14H16O3
59780
DL-Kavain, analytical standard
C14H14O3