EMAIL THIS PAGE TO A FRIEND
Acta dermato-venereologica
"Extra-edge effect": a further problem in patch test diagnosis?
PMID 18311466
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21620 Disperse Blue 124, for microscopy