EMAIL THIS PAGE TO A FRIEND
Cancer Research
Clonal selection in malignant transformation of human fibroblasts transduced with defined cellular oncogenes.
PMID 18316605
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-516
Anti-Ras Antibody, clone RAS10, clone RAS10, Upstate®, from mouse