EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Nanoparticle: is it promising in capillary electrophoresis?
PMID 18317740
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
662534
4-Mercaptobenzoic acid, technical grade, 90%
C7H6O2S
706329
4-Mercaptobenzoic acid, 99%
C7H6O2S