EMAIL THIS PAGE TO A FRIEND
The journal of medical investigation : JMI
Problems in three Japanese drug users with Human Immunodeficiency Virus infection.
PMID 18319560
Abstract
Numbers of individuals infected with Human Immunodeficiency Virus (HIV) are increasing in Japan. The majority of them are Men who have sex with men and a part of them take drugs as 'Sex drug' at their sexual intercourse. Especially, Amyl nitrite, Methamphetamine, 5-methoxy-N, N-diisopropyltryptamine (5-MeO-DIPT; Foxy), and 3, 4-methylenedioxy- methamphetamine (MDMA; Ecstasy) are used, and they sometimes cause the physical and mental disorders. However, the actual drug inducing troubles among Japanese HIV-infected drug users had not yet been discussed enough. In this report, we describe three cases with HIV infection; a case developed severe neuroleptic malignant syndrome (NMS) after taking 5-MeO-DIPT, a case with persistent convulsion due to multiple drug intake and a case with rhabdomyolysis due to the non-subjective methamphetamine intake. Through these cases, we raise and discuss several underlying problems associated with drug use among HIV-infected individuals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
286583
5-Methoxytryptamine, 97%
C11H14N2O