EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Determining the levels of benazol II in environmental objects].
PMID 1832128
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
533203
2-(2-Hydroxy-5-methylphenyl)benzotriazole, 97%
C13H11N3O