EMAIL THIS PAGE TO A FRIEND
Ugeskrift for laeger
[Serious debate on unconventional treatment, thank you].
PMID 18326084
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5775
Bendroflumethiazide, analytical standard
C15H14F3N3O4S2