EMAIL THIS PAGE TO A FRIEND
Plant physiology
Construction of a functional CMP-sialic acid biosynthesis pathway in Arabidopsis.
PMID 18326787
Abstract
Previous studies have reported that plants contain negligible amounts of free or protein-bound N-acetylneuraminic acid (Neu5Ac). This is a major disadvantage for the use of plants as a biopharmaceutical expression system, since N-glycans with terminal Neu5Ac residues are important for the biological activities and half-lives of recombinant therapeutic glycoproteins in humans. For the synthesis of Neu5Ac-containing N-glycans, plants have to acquire the ability to synthesize Neu5Ac and its nucleotide-activated derivative, cytidine monophospho-N-acetylneuraminic acid. In this study, we have generated transgenic Arabidopsis (Arabidopsis thaliana) plants expressing three key enzymes of the mammalian Neu5Ac biosynthesis pathway: UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase, N-acetylneuraminic acid phosphate synthase, and CMP-N-acetylneuraminic acid synthetase. Simultaneous expression of UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase and N-acetylneuraminic acid phosphate synthase resulted in the generation of significant Neu5Ac amounts (1,275 nmol g(-1) fresh weight in leaves) in planta, which could be further converted to cytidine monophospho-N-acetylneuraminic acid (2.4 nmol g(-1) fresh weight in leaves) by coexpression of CMP-N-acetylneuraminic acid synthetase. These findings are a major step toward the production of Neu5Ac-containing glycoproteins in plants.