EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Simple and facile synthesis of tetralone-spiro-glutarimides and spiro-bisglutarimides from Baylis-Hillman acetates.
PMID 18327328
Abstract
A simple and convenient synthesis of di(E)-arylidene-tetralone-spiro-glutarimides from Baylis-Hillman acetates via an interesting biscyclization strategy involving facile C-C and C-N bond formation is described. Also, one-pot multistep transformation of the Baylis-Hillman acetates into di(E)-arylidene-spiro-bisglutarimides is presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
178098
Glutarimide, 98%
C5H7NO2