EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Palladium-catalyzed synthesis of indene derivatives via propargylic carbonates with in situ generated organozinc compounds.
PMID 18327329
Abstract
The palladium-catalyzed carboannulation and arylation reaction of propargylic carbonates with in situ generated organozinc compounds produced an important new class of indene derivatives. The reaction proceeded under mild conditions, and indene products were isolated in good to excellent yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
130737
3-Iodotoluene, 99%
C7H7I