EMAIL THIS PAGE TO A FRIEND
Epilepsia
Clinical pharmacology of parenteral use of antiepileptic drugs.
PMID 18329998
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0117
Fosphenytoin disodium salt hydrate, ≥98% (HPLC)
C16H13N2Na2O6P · xH2O