EMAIL THIS PAGE TO A FRIEND
Journal of economic entomology
Pymetrozine causes a nontarget pest, the Colorado potato beetle (Coleoptera: Chrysomelidae), to leave potato plants.
PMID 18330119
Abstract
Pymetrozine is a selective insecticide that targets aphids. Published assessments of the effects of pymetrozine on nontarget organisms focus mainly on predatory insects, and they rarely indicate toxicity. In a laboratory bioassay, survival of Colorado potato beetle, Leptinotarsa decemlineata (Say) (Coleoptera: Chrysomelidae), larvae was not affected by pymetrozine exposure. We subsequently used pymetrozine to implement low-aphid-density treatments in a field experiment that involved separate manipulations of Colorado potato beetle density. Unexpectedly, the addition of Colorado potato beetle adults and eggs did not increase the densities of Colorado potato beetle larvae in plots that were sprayed with pymetrozine (applied with water and an adjuvant). In control plots sprayed with water and adjuvant (without pymetrozine), addition of Colorado potato beetles increased densities of their larvae. Data collected on a smaller scale suggest that a behavioral mechanism underlies the population-level pattern: Colorado potato beetle larvae become more active and are less likely to remain on a host plant after exposure to pymetrozine. Thus, potato, Solanum tuberosum L., growers who use pymetrozine against aphids also might benefit in terms of Colorado potato beetle control.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46119
Pymetrozin, PESTANAL®, analytical standard
C10H11N5O