EMAIL THIS PAGE TO A FRIEND
Developmental Dynamics
Differences in early lineage segregation between mammals.
PMID 18330925
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB4123 Anti-Cdx2 Antibody