EMAIL THIS PAGE TO A FRIEND
World Journal of Gastroenterology
Synergistic growth inhibitory effects of Phyllanthus emblica and Terminalia bellerica extracts with conventional cytotoxic agents: doxorubicin and cisplatin against human hepatocellular carcinoma and lung cancer cells.
PMID 18330936
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31525
TCA, PESTANAL®, analytical standard
C2Cl3NaO2