EMAIL THIS PAGE TO A FRIEND
Journal of microscopy
SNOM images of X-ray radiographs at nano-scale stored in a thin layer of lithium fluoride.
PMID 18331500
Abstract
In this work, we report a method to observe soft X-ray radiographs at nanoscale of various kind of samples, biological and metallic, stored in a thin layer of lithium fluoride, employing scanning near-field optical microscopy with an optical resolution that reaches 50 nm. Lithium fluoride material works as a novel image detector for X-ray nano-radiographs, due to the fact that extreme ultraviolet radiation and soft X-rays efficiently produce stable point defects emitting optically stimulated visible luminescence in a thin surface layer. The bi-dimensional distribution of the so-created defects depends on the local nanostructure of the investigated sample.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
62497
Lithium fluoride, BioUltra, ≥99.0% (F)
FLi