EMAIL THIS PAGE TO A FRIEND
Pediatric blood & cancer
Central neurocytoma responsive to topotecan, ifosfamide, carboplatin.
PMID 18338396
Abstract
A 5-year-old male presented with spinal cord drop metastasis from a recurrent neurocytoma. Topotecan (0.5 mg/m(2)) and carboplatin (250 mg/m(2)) were administered on days 1-3 and ifosfamide (1,800 mg/m(2)) on days 1-5, every 21 days, for three cycles and resulted in complete response without severe complications. A literature review yielded 20 patients with central neurocytoma but no complete responses. The complete response of central neurocytoma to chemotherapy only reported here should be helpful to those caring for patients with this rare tumor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I4909
Ifosfamide, ≥98%
C7H15Cl2N2O2P
I0060000
Ifosfamide, European Pharmacopoeia (EP) Reference Standard
C7H15Cl2N2O2P