EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Inhibitory effects of emodin on proliferation of rat vascular smooth muscle cell induced by angiotensin II].
PMID 18338623
Abstract
To investigate the effects of emodin on the proliferation of cultured rat vascular smooth muscle cell (VSMC) induced by angiotensin II. VSMCs were cultured by explant method. Cell proliferation model was established by stimulation with Ang II. Cell proliferation was measured by MTT assay to observe the effects of emodin (10, 20, 40 and 80 micromol x L(-1)) and N(G)-nitro-L-arginine methyl ester (L-NAME, 100 micromol x L(-1)) on VSMC proliferation induced by Ang II. The expression of PCNA was measured by immunohistochemical staining. Nitric oxide (NO) level was measured by Griess reagent. Nitric oxide synthase (NOS) and inducible nitric oxide synthase (iNOS) levels were detected by chemical colorimetric method. mRNA expression of iNOS was measured by reverse transcription polymerase chain reaction (RT-PCR). Emodin at the doses range from 10 to 80 mol x L(-1) inhibited cell proliferation in a dose and time-dependent manner. The inhibitory effects were partly blocked by 100 mol x L(-1) of L-NAME. Emodin markedly decreased the expression of PCNA in VSMC, increased NO, NOS and iNOS levels, and increased iNOS mRNA expression in VSMC. Emodin could inhibite VSMCs proliferation induced by Ang II. Inhibiting the expression of PCNA, increasing the NO secretion and upregulating the iNOS gene expression might be associated with the inhibitory effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11030
L-Arginine methyl ester dihydrochloride, ≥98.0% (AT)
C7H16N4O2 · 2HCl