EMAIL THIS PAGE TO A FRIEND
Chemistry and physics of lipids
Monoepoxy octadecadienoates and monoepoxy octadecatrienoates 2: mass spectral characterization.
PMID 18339313
Abstract
Methyl esters of C18 polyunsaturated fatty acids, including gamma-linolenic acid, alpha-linolenic acid and stearidonic acid, were epoxidised using m-chloroperbenzoic acid. Nine monoepoxides were obtained by normal-phase HPLC, identified using LC-MS and NMR, and characterized by NMR spectroscopy and mass spectrometry. This study is focused on structural characterization using LC-MS and LC/APCI/MS/MS. The elution profiles of these monoepoxides in RP-HPLC are determined as 12,13->9,10->6,7-epoxy, 9,10->15,16->12,13-epoxy and 15,16->12,13->9,10-epoxy derivatives of gamma-linolenic, alpha-linolenic and stearidonic acid methyl esters, respectively. The major diagnostic fragmentations in MS/MS identified are postulated to be induced by cleavages of the epoxide ring and alpha-bond cleavage to the epoxy group from [M+H]+ and/or [M+H-MeOH]+.