EMAIL THIS PAGE TO A FRIEND
Xenobiotica; the fate of foreign compounds in biological systems
Dietary salt does not influence the disposition of verapamil enantiomers in relation to efflux transporter ABCB1 genetic polymorphism in healthy Korean subjects.
PMID 18340565
Abstract
To evaluate the effects of dietary salt on the stereoselective disposition of verapamil enantiomers in relation to the transporter ABCB1 2677GG/3435CC and 2677TT/3435TT haplotypes, ten healthy subjects were asked to take diets of three different salt levels for 7 days in a randomized, three-way crossover manner. The plasma concentrations of verapamil and norverapamil enantiomers were determined after a single oral dose of 240 mg verapamil on the last day of each phase. Pharmacokinetic parameters were calculated by non-compartmental analysis techniques and compared among the three different dietary salt phases. Compared with the medium salt diet, the high and low salt diets had no significant effect on the disposition of verapamil enantiomers. Moreover, the ABCB1 haplotypes did not alter the impact of dietary salt, although ABCB1 2677TT/3435TT subjects had slightly, but not significantly, higher C(max) and area under the curve (AUC) and lower T(max) for the verapamil enantiomers than did 2677GG/3435CC subjects in each salt phase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V108
(±)-Norverapamil hydrochloride, ≥98% (HPLC)
C26H36N2O4 · HCl