EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Constrained analogues of tocainide as potent skeletal muscle sodium channel blockers towards the development of antimyotonic agents.
PMID 18342401
Abstract
1-Benzyl-N-(2,6-dimethylphenyl)piperidine-3-carboxamide and 4-benzyl-N-(2,6-dimethylphenyl)piperazine-2-carboxamide, two conformationally restricted analogues of tocainide, were designed and synthesized as voltage-gated skeletal muscle sodium channel blockers. They showed, with respect to tocainide, a marked increase in both potency and use-dependent block.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0202
Tocainide hydrochloride, ≥98% (HPLC), solid
C11H16N2O · HCl